2,3-Dibromonaphthalene 98%

Supplier: Ambeed
Warning

Synonyms: 2,3-Diiodonaphthalene

A127301-100MG A127301-10G A127301-250MG A127301-1G A127301-25G A127301-5G
103800-946EA 26 USD
103800-946 76797-712 76797-714 76797-716 77313-194 77313-196 77313-198
2,3-Dibromonaphthalene 98%
2,3-Dibromonaphthalene

Precaución: for Research Use Only

Formula: C₁₀H₆Br₂
MW: 285.97
Boiling Pt: 341.7 °C (760 mm Hg)
Flash Pt: 186.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00021589
CAS Number: 13214-70-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR