2,3-Dibromo-4-methylpyridine 97%

Supplier: Ambeed
Warning

A107182-250MG A107182-25G A107182-100G A107182-10G
103797-148EA 18 USD
103797-148 103822-910 77135-762 77592-148 77684-966 77684-967
2,3-Dibromo-4-methylpyridine 97%
2,3-Dibromo-4-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅Br₂N
MW: 250.91 g/mol
Boiling Pt: 270.4 °C (760 mm Hg)
Flash Pt: 117.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09753264
CAS Number: 871483-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR