2,3-Diaminopyridine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Pyridine-2,3-diyldiamine
A218774-1G
A218774-5G
A218774-100G
A218774-10G
A218774-250MG
A218774-25G
103818-748EA
9
USD
103818-748
103849-618
77088-150
77088-152
77275-430
77275-432
77275-434
77303-514
2,3-Diaminopyridine 97%
2,3-Diaminopyridine
Precaución: for Research Use Only
Formula:
C₅H₇N₃ MW: 109.13 g/mol Boiling Pt: 311.3 °C (760 mm Hg) Melting Pt: 110…115 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006319 CAS Number: 452-58-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...