2,3-Diaminopyrazine 97%

Supplier: Ambeed
Warning

Synonyms: Pyrazine-2,3-diamine

A145988-100MG A145988-250MG A145988-25G A145988-5G A145988-1G
103803-612EA 19 USD
103803-612 103841-698 77247-120 77247-122 77299-486 77299-488 77317-006
2,3-Diaminopyrazine 97%
2,3-Diaminopyrazine

Precaución: for Research Use Only

Formula: C₄H₆N₄
MW: 110.1172 g/mol
Boiling Pt: 351.6 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD07778385
CAS Number: 13134-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR