2,3-Diaminonaphthalene 97%

Supplier: Ambeed
Warning

Synonyms: 2,3-Naphthalenediamine, DAN, 2,3-naphthylenediamine

A100422-10G A100422-1MG A100422-250MG A100422-25G A100422-1G A100422-5G A100422-100G
103795-430EA 20 USD
103795-430 103821-126 76686-982 76686-984 76686-986 76686-988 77296-678 77296-680 77623-806
2,3-Diaminonaphthalene 97%
2,3-Diaminonaphthalene

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂
MW: 158.2 g/mol
Melting Pt: 198…200 °C
Flash Pt: 212.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004116
CAS Number: 771-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR