2,3,5-Trimethylphenol 98%

Supplier: Ambeed
Danger

A450650-1000G A450650-25G A450650-100G A450650-10G A450650-500G
103810-356EA 172 USD
103810-356 76842-676 76842-678 77252-774 77252-776 77252-778
2,3,5-Trimethylphenol 98%
2,3,5-Trimethylphenol

Precaución: for Research Use Only

Formula: C₉H₁₂O
MW: 136.19 g/mol
Boiling Pt: 231.1 °C (760 mm Hg)
Melting Pt: 92…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00002228
CAS Number: 697-82-5
UN: 2430
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR