2,3,5-Trichlorophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2,3,5-Trichlorobenzeneboronic acid, Dihydroxy(2,3,5-trichlorophenyl)borane, (2,3,5-Trichlorophenyl)metaboric acid

A178759-25G A178759-5G A178759-250MG A178759-10G
103807-366EA 15 USD
103807-366 103845-706 77083-144 77083-146 77151-562 77685-403
2,3,5-Trichlorophenylboronic acid 98%
2,3,5-Trichlorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₄BCl₃O₂
MW: 225.26 g/mol
Boiling Pt: 383.4 °C (760 mm Hg)
Melting Pt: 245 °C
Flash Pt: 185.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01630856
CAS Number: 212779-19-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR