2,3,5-Trichloroisonicotinic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2,3,5-Trichloropyridine-4-carboxylic acid

A257719-10G A257719-1G
103812-334EA 7 USD
103812-334 103852-202 76786-914 77304-718
2,3,5-Trichloroisonicotinic acid 95%
2,3,5-Trichloroisonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₂Cl₃NO₂
MW: 226.44 g/mol
Melting Pt: 193…194 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11044390
CAS Number: 406676-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR