2,3,5,6-Tetrafluorophenol 95%

Supplier: Ambeed
Warning

A845946-25G A845946-5G A845946-10G A845946-1G A845946-500G A845946-100G
103841-220EA 13 USD
103841-220 76845-466 76845-468 77256-262 77256-264 77256-266 77569-574
2,3,5,6-Tetrafluorophenol 95%
2,3,5,6-Tetrafluorophenol

Precaución: For research use only.

Formula: C₆H₂F₄O
Boiling Pt: 140.2±35.0 °C (760 mm Hg)
Melting Pt: 37…39 °C
Flash Pt: 79.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00002157
CAS Number: 769-39-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR