2,3,5,6-Tetrafluorobenzoic acid 98%

Supplier: Ambeed
Danger

A558756-10G A558756-500G A558756-25G A558756-5G A558756-1G
103828-738EA 6 USD
103828-738 103839-868 76774-896 76774-898 76843-968 76843-970 77307-806
2,3,5,6-Tetrafluorobenzoic acid 98%
2,3,5,6-Tetrafluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₂F₄O₂
MW: 194.09 g/mol
Boiling Pt: 227.9 °C (760 mm Hg)
Melting Pt: 150…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00002408
CAS Number: 652-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR