2,3,5,6-Tetrafluoro-1,4-benzenedimethanol 97%

Supplier: Ambeed
Warning

Synonyms: 2,3,5,6-Tetrafluoro-1,4-bis(hydroxymethyl)benzene, 2,3,5,6-Tetrafluoro-p-xylene-alpha,alpha'-diol, 2,3,5,6-Tetrafluoro-alpha,alpha'-dihydroxy-p-xylene

A114783-5G A114783-25G
103798-472EA 5 USD
103798-472 103824-314 77297-324 77882-947
2,3,5,6-Tetrafluoro-1,4-benzenedimethanol 97%
2,3,5,6-Tetrafluoro-1,4-benzenedimethanol

Precaución: for Research Use Only

Formula: C₈H₆F₄O₂
Boiling Pt: 256 °C (760 mm Hg)
Melting Pt: 129 °C (760 mm Hg)
Flash Pt: 108.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00229147
CAS Number: 92339-07-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR