2,3,4-Trimethoxybenzoic acid 97%

Supplier: Ambeed
Warning

A313262-100G A313262-10G A313262-500G A313262-5G
103810-078EA 382 USD
103810-078 103835-908 76693-636 76693-638 76693-640 77306-080
2,3,4-Trimethoxybenzoic acid 97%
2,3,4-Trimethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₅
MW: 212.2 g/mol
Boiling Pt: 321 °C (760 mm Hg)
Melting Pt: 99…102 °C
Flash Pt: 123.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00002433
CAS Number: 573-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR