2,3,4-Trihydroxybenzoic acid 99%

Supplier: Ambeed
Warning

A101419-100G A101419-10G A101419-25G A101419-5G
103821-652EA 6 USD
103821-652 77135-482 77135-484 77135-486 77297-102
2,3,4-Trihydroxybenzoic acid 99%
2,3,4-Trihydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₆O₅
MW: 170.12 g/mol
Boiling Pt: 437.5 °C (760 mm Hg)
Melting Pt: 205 °C
Storage Temperature: Ambient
MDL Number: MFCD00002447
CAS Number: 610-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR