2,3,4-Trihydroxybenzaldehyde 98%
Supplier: Ambeed
|
Warning
|
A216246-100G
A216246-10G
A216246-1G
A216246-25G
103849-258EA
7
USD
103849-258
77249-232
77249-234
77249-236
77303-430
2,3,4-Trihydroxybenzaldehyde 98%
2,3,4-Trihydroxybenzaldehyde
Precaución: for Research Use Only
Formula:
C₇H₆O₄ MW: 154.12 g/mol Boiling Pt: 301.9 °C (760 mm Hg) Melting Pt: 159…161 °C Flash Pt: 112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003325 CAS Number: 2144-08-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...