2,3,4,6-Tetrachloropyridine 97%

Supplier: Ambeed
Warning

A291233-100MG A291233-250MG A291233-1G A291233-5G
103799-576EA 14 USD
103799-576 103813-902 103835-042 77141-332 77141-334 77305-604 77305-606
2,3,4,6-Tetrachloropyridine 97%
2,3,4,6-Tetrachloropyridine

Precaución: for Research Use Only

Formula: C₅HCl₄N
MW: 216.88 g/mol
Boiling Pt: 108…112 °C (12 mm Hg)
Melting Pt: 68…70 °C
Flash Pt: 135.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01312169
CAS Number: 14121-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR