1,8-Diazabicyclo[5.4.0]undec-7-ene 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
2,3,4,6,7,8,9,10-Octahydropyrimido[1,2-a]azepine, DBU
A669250-100G
A669250-1G
A669250-25G
A669250-5G
103810-506EA
42
USD
103810-506
103840-488
76819-862
76819-864
76819-866
76844-694
77552-034
1,8-Diazabicyclo[5.4.0]undec-7-ene 98%
1,8-Diazabicyclo[5.4.0]undec-7-ene
Precaución: for Research Use Only
Formula:
C₉H₁₆N₂ MW: 152.24 g/mol Boiling Pt: 274.6 °C (760 mm Hg) Melting Pt: 70 °C Flash Pt: 116 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006930 CAS Number: 6674-22-2 UN: 2922 ADR: 8,6.1 Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...