2,3,4,5-Tetramethyl-2-cyclopentenone 95%

Supplier: Ambeed
Warning

Synonyms: 2,3,4,5-Tetramethylcyclopent-2-enone

A173061-100G A173061-10G A173061-500G A173061-1G A173061-25G
103806-786EA 5 USD
103806-786 103845-080 76813-278 76813-280 76813-282 77082-302 77301-244
2,3,4,5-Tetramethyl-2-cyclopentenone 95%
2,3,4,5-Tetramethyl-2-cyclopentenone

Precaución: for Research Use Only

Formula: C₉H₁₄O
MW: 138.21 g/mol
Boiling Pt: 196 °C (760 mm Hg)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00010248
CAS Number: 54458-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR