Pentafluorophenol 98%

Supplier: Ambeed
Warning

Synonyms: 2,3,4,5,6-Pentafluorophenol

A112045-100G A112045-10G A112045-25G A112045-500G A112045-5G
103823-768EA 12 USD
103823-768 76718-126 76718-128 76718-130 76718-132 77284-204
Pentafluorophenol 98%
Pentafluorophenol

Precaución: for Research Use Only

Formula: C₆HF₅O
MW: 184.07 g/mol
Boiling Pt: 146.3 °C (760 mm Hg)
Melting Pt: 34…36 °C
Flash Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00002156
CAS Number: 771-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR