2,2′-sulfonyldiethanol 65% in water

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxyethyl sulphone, bis(β-Hydroxyethyl)sulphone

A310335-5G A310335-100G A310335-10G A310335-25G A310335-500G
103835-798EA 7 USD
103835-798 76838-576 77250-952 77250-954 77250-956 77593-862
2,2′-sulfonyldiethanol 65% in water
2,2′-sulfonyldiethanol

Precaución: for Research Use Only

Formula: O₂S(CH₂CH₂OH)₂
MW: 154.18 g/mol
Boiling Pt: 432.3 °C (760 mm Hg)
Flash Pt: 215.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00002907
CAS Number: 2580-77-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR