2,2-Dimethylpyrrolidine hydrochloride 95%

Supplier: Ambeed
Warning

A180979-250MG A180979-1G A180979-5G
103807-576EA 21 USD
103807-576 103845-938 77029-432 77083-452 77083-454
2,2-Dimethylpyrrolidine hydrochloride 95%
2,2-Dimethylpyrrolidine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄ClN
Melting Pt: 88.8…90.4 °C
Storage Temperature: Ambient
MDL Number: MFCD09759233
CAS Number: 623580-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR