2,2-Dimethyl-4-pentenoic acid 98%

Supplier: Ambeed
Danger

Synonyms: 2,2-Dimethylpent-4-enoic acid

A549032-1G A549032-5G A549032-10G A549032-250MG A549032-25G
103828-682EA 23 USD
103828-682 103839-804 76843-880 76843-882 77253-796 77253-798 77253-800
2,2-Dimethyl-4-pentenoic acid 98%
2,2-Dimethyl-4-pentenoic acid

Precaución: for Research Use Only

Formula: C₇H₁₂O₂
Boiling Pt: 202.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00012304
CAS Number: 16386-93-9
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR