2,2-Dimethylmorpholine 97%

Supplier: Ambeed
Warning

A103128-10G A103128-250MG A103128-1G A103128-5G A103128-25G
103796-486EA 35 USD
103796-486 103822-190 77025-250 77025-252 77283-044 77309-064 77670-488
2,2-Dimethylmorpholine 97%
2,2-Dimethylmorpholine

Precaución: for Research Use Only

Formula: C₆H₁₃NO
Boiling Pt: 144.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09701959
CAS Number: 147688-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR