t-Boc-N-Amido-PEG2-CH2CO2H 97%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: Boc-NH-PEG2-CH2COOH, 2,2-Dimethyl-4-oxo-3,8,11-trioxa-5-azatridecan-13-oic acid

A167750-100MG A167750-250MG A167750-1G A167750-5G A167750-10G
103806-216EA 22 USD
103806-216 103844-476 76985-296 76985-298 77081-508 77081-510 77683-375
t-Boc-N-Amido-PEG2-CH2CO2H 97%, Ambeed.Inc
t-Boc-N-Amido-PEG2-CH2CO2H

Precaución: For research use only.

Formula: C₁₁H₂₁NO₆
MW: 263.288 g/mol
Boiling Pt: 630.4 °C (760 mm Hg)
Melting Pt: 240 °C
Storage Temperature: Freezer
MDL Number: MFCD17019374
CAS Number: 108466-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR