(Boc-amino)-PEG4-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Boc-NH-PEG4-CH2CH2COOH, Carboxy-PEG4-(Boc-amine), 15-[(tert-Butoxycarbonyl)amino]-4,7,10,13-tetraoxapentadecanoic acid, 2,2-Dimethyl-4-oxo-3,8,11,14,17-pentaoxa-5-azaicosan-20-oic acid, 15-(Boc-amino)-4,7,10,13-tetraoxapentadecanoic acid
A108006-5G
A108006-250MG
A108006-100MG
A108006-25G
103797-276EA
32
USD
103797-276
103823-052
77309-892
77549-090
77610-850
77670-540
(Boc-amino)-PEG4-carboxylic acid 97%
(Boc-amino)-PEG4-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₆H₃₁NO₈ MW: 365.42 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD07781254 CAS Number: 756525-91-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...