2,2-Dimethyl-3-phenylpropanoic acid 98%

Supplier: Ambeed
Warning

A141975-100MG A141975-1G
103803-086EA 104 USD
103803-086 76996-898 77316-288
2,2-Dimethyl-3-phenylpropanoic acid 98%
2,2-Dimethyl-3-phenylpropanoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
MW: 178.22 g/mol
Boiling Pt: 288.3 °C (760 mm Hg)
Flash Pt: 185.4 °C
Storage Temperature: Ambient
MDL Number: MFCD01722147
CAS Number: 5669-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR