2,2-Dimethyl-3-oxobutanenitrile 97%

Supplier: Ambeed
Danger

A101269-100MG A101269-250MG
103795-884EA 219 USD
103795-884 103821-570 76809-234 76809-236
2,2-Dimethyl-3-oxobutanenitrile 97%
2,2-Dimethyl-3-oxobutanenitrile

Precaución: for Research Use Only

Formula: C₆H₉NO
MW: 111.14 g/mol
Boiling Pt: 152.2±13.0 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD18827245
CAS Number: 37719-02-1
UN: 3273
ADR: 3,6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR