2,2-Dimethyl-1,3-dioxane-4,6-dione (isopropylidene malonate, meldrum's acid) 98%
Supplier: Ambeed
|
Danger
|
A412608-25G
A412608-100G
A412608-500G
103810-304EA
53
USD
103810-304
76841-840
77551-234
77594-268
2,2-Dimethyl-1,3-dioxane-4,6-dione (isopropylidene malonate, meldrum's acid) 98%
2,2-Dimethyl-1,3-dioxane-4,6-dione (isopropylidene malonate, meldrum's acid)
Precaución: for Research Use Only
Formula:
C₆H₈O₄ Boiling Pt: 356.7 °C (760 mm Hg) Melting Pt: 92…96 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006638 CAS Number: 2033-24-1 UN: 3077 ADR: 9,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...