2,2'-Dimethyl-[1,1'-biphenyl]-4-carboxylic acid 98%

Supplier: Ambeed
Warning

A154238-100MG A154238-250MG A154238-1G
103804-616EA 187 USD
103804-616 103842-800 76823-914 76823-916 77318-502
2,2'-Dimethyl-[1,1'-biphenyl]-4-carboxylic acid 98%
2,2'-Dimethyl-[1,1'-biphenyl]-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD18318906
CAS Number: 69535-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR