2,2-Dimethoxypropan-1-amine 95%
Supplier: Ambeed
|
Warning
|
A194561-1G
A194561-5G
A194561-100MG
A194561-10G
A194561-250MG
103816-666EA
23
USD
103816-666
103847-276
77085-200
77085-202
77237-008
77237-010
77237-012
2,2-Dimethoxypropan-1-amine 95%
2,2-Dimethoxypropan-1-amine
Precaución: for Research Use Only
Formula:
C₅H₁₃NO₂ Boiling Pt: 146 °C at 751 mmHg Storage Temperature: Refrigerator |
MDL Number:
MFCD10698200 CAS Number: 131713-50-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...