N-(3-Pyridyl)pivalamide 95%

Supplier: Ambeed
Warning

Synonyms: 2,2-Dimethyl-N-pyridin-3-yl-propionamide, 2,2-Dimehtyl-N-pyridin-3-yl-propionamide

A146320-1G A146320-25G A146320-5G
103803-654EA 8 USD
103803-654 103841-740 77317-062 77317-064 77317-066
N-(3-Pyridyl)pivalamide 95%
N-(3-Pyridyl)pivalamide

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₂O
MW: 178.23 g/mol
Boiling Pt: 337.5 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00996243
CAS Number: 70298-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR