2,2-Difluoropropylamine hydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Amino-2,2-difluoropropane hydrochloride, 2,2-Difluoropropan-1-amine hydrochloride
A133491-100MG
A133491-250MG
A133491-1G
103801-880EA
33
USD
103801-880
77246-640
77246-642
77314-532
2,2-Difluoropropylamine hydrochloride 97%
2,2-Difluoropropylamine hydrochloride
Precaución: for Research Use Only
Formula:
C₃H₇F₂N·ClH MW: 131.552 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD06659051 CAS Number: 868241-48-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...