2,2-Difluoro-1,3-benzodioxole-5-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2,2-Difluorobenzo[d][1,3]dioxole-5-carboxylic acid

A163768-100G A163768-10G A163768-1G A163768-25G
103805-762EA 5 USD
103805-762 103844-024 77247-726 77247-728 77320-134 77320-136
2,2-Difluoro-1,3-benzodioxole-5-carboxylic acid 98%
2,2-Difluoro-1,3-benzodioxole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₄F₂O₄
MW: 202.12 g/mol
Boiling Pt: 266.1 °C (760 mm Hg)
Melting Pt: 178 °C
Flash Pt: 114.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00792417
CAS Number: 656-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR