2,2-Difluoro-1,3-benzodioxole-5-carboxaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 2,2-Difluorobenzo[d][1,3]dioxole-5-carbaldehyde

A377087-1G A377087-5G A377087-10G A377087-25G
103826-600EA 6 USD
103826-600 103837-628 76840-932 76840-934 77195-556 77306-890
2,2-Difluoro-1,3-benzodioxole-5-carboxaldehyde 97%
2,2-Difluoro-1,3-benzodioxole-5-carboxaldehyde

Precaución: for Research Use Only

Formula: C₈H₄F₂O₃
MW: 186.11 g/mol
Boiling Pt: 202…203 °C (760 mm Hg)
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00792420
CAS Number: 656-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR