5-Amino-2,2-difluoro-1,3-benzodioxole 97%

Supplier: Ambeed
Warning

A122948-10G A122948-1G A122948-5G A122948-100G A122948-25G
103800-312EA 6 USD
103800-312 77136-590 77312-430 77312-432 77549-446 77592-396
5-Amino-2,2-difluoro-1,3-benzodioxole 97%
5-Amino-2,2-difluoro-1,3-benzodioxole

Precaución: for Research Use Only

Formula: C₇H₅F₂NO₂
Boiling Pt: 259.6 °C (760 mm Hg)
Flash Pt: 110.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00190144
CAS Number: 1544-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR