Glycolaldehyde diethyl acetal 97%

Supplier: Ambeed
Warning

Synonyms: Hydroxyacetaldehyde diethyl acetal, 2,2-Diethoxyethanol

A120630-10G A120630-25G A120630-500G A120630-1G A120630-5G
103799-974EA 6 USD
103799-974 77108-570 77108-572 77108-574 77312-082 77312-084
Glycolaldehyde diethyl acetal 97%
Glycolaldehyde diethyl acetal

Precaución: for Research Use Only

Formula: HOCH₂CH(OCH₂CH₃)₂
MW: 134.18 g/mol
Boiling Pt: 166…167 °C (760 mm Hg)
Flash Pt: 62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00051486
CAS Number: 621-63-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR