2,2'-Bithiophene 98%

Supplier: Ambeed
Warning

Synonyms: 2,2′-Dithienyl, 2,2′-Bithienyl

A112167-100G A112167-10G A112167-1G A112167-25G A112167-500G A112167-5G
103823-802EA 10 USD
103823-802 77245-740 77245-742 77245-744 77245-746 77245-748 77310-612
2,2'-Bithiophene 98%
2,2'-Bithiophene

Precaución: for Research Use Only

Formula: C₈H₆S₂
MW: 166.27 g/mol
Boiling Pt: 260 °C (760 mm Hg)
Melting Pt: 32…33 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005414
CAS Number: 492-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR