2,2-Bis[4-(4-aminophenoxy)phenyl]hexafluoropropane 98%

Supplier: Ambeed
Warning

Synonyms: 4,4'-(Hexafluoroisopropylidene)bis[(4-aminophenoxy)benzene]

A101753-100G A101753-25G A101753-500G A101753-5G
103796-116EA 7 USD
103796-116 103821-802 77093-520 77093-522 77093-524 77282-822
2,2-Bis[4-(4-aminophenoxy)phenyl]hexafluoropropane 98%
2,2-Bis[4-(4-aminophenoxy)phenyl]hexafluoropropane

Precaución: for Research Use Only

Formula: C₂₇H₂₀F₆N₂O₂
MW: 518.46 g/mol
Boiling Pt: 544.8 °C (760 mm Hg)
Melting Pt: 162…165 °C
Flash Pt: 283.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00015723
CAS Number: 69563-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR