2,2'-Bis(trifluoromethyl)benzidine 98%

Supplier: Ambeed
Warning

Synonyms: 2,2'-Bis(trifluoromethyl)-[1,1'-biphenyl]-4,4'-diamine, 4,4'-Diamino-2,2'-bis(trifluoromethyl)biphenyl

A150373-100G A150373-10G A150373-500G A150373-1G A150373-25G A150373-5G
103804-154EA 5 USD
103804-154 103842-288 77247-220 77247-222 77247-224 77299-794 77317-804 77317-806
2,2'-Bis(trifluoromethyl)benzidine 98%
2,2'-Bis(trifluoromethyl)benzidine

Precaución: for Research Use Only

Formula: C₁₄H₁₀F₆N₂
Boiling Pt: 376.9 °C (760 mm Hg)
Melting Pt: 183 °C
Flash Pt: 171.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00190155
CAS Number: 341-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR