2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl 97%

Supplier: Ambeed
Warning

Synonyms: racemic-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, rac-BINAP, (+/-)-(1,1'-Binaphthalene-2,2'-diyl)bis(diphenylphosphine), (±)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, (+/-)-BINAP

A122779-100G A122779-10G A122779-25G A122779-500G A122779-1G A122779-5G
103800-280EA 5 USD
103800-280 76822-522 76822-524 76822-526 76822-528 77297-894 77312-398
2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl 97%
2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl

Precaución: for Research Use Only

Formula: C₄₄H₃₂P₂
MW: 622.67 g/mol
Boiling Pt: 724.3 °C (760 mm Hg)
Melting Pt: 283…286 °C
Flash Pt: 419 °C
Storage Temperature: Ambient
MDL Number: MFCD00010805
CAS Number: 98327-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR