2,2-Bis(((2-cyano-3,3-diphenylacryloyl)oxy)methyl)propane-1,3-diyl bis(2-cyano-3,3-diphenylacrylate) 95%
Supplier: Ambeed
|
Warning
|
A239093-100G
A239093-10G
A239093-1G
A239093-25G
A239093-5G
103811-264EA
10
USD
103811-264
103851-062
76826-686
76826-688
76826-690
77304-134
77304-136
77304-138
2,2-Bis(((2-cyano-3,3-diphenylacryloyl)oxy)methyl)propane-1,3-diyl bis(2-cyano-3,3-diphenylacrylate) 95%
2,2-Bis(((2-cyano-3,3-diphenylacryloyl)oxy)methyl)propane-1,3-diyl bis(2-cyano-3,3-diphenylacrylate)
Precaución: for Research Use Only
Formula:
C₆₉H₄₈N₄O₈ Boiling Pt: 1077.4±65.0 °C (760 mm Hg) Flash Pt: 418.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD21607536 CAS Number: 178671-58-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...