2,2'-Biquinoline 98%

Supplier: Ambeed
Warning

A356507-1G A356507-25G A356507-5G A356507-10G A356507-100G
103826-172EA 14 USD
103826-172 103837-180 76840-316 76840-318 76840-320 77033-152 77872-540
2,2'-Biquinoline 98%
2,2'-Biquinoline

Precaución: for Research Use Only

Formula: C₁₈H₁₂N₂
Melting Pt: 192…195 °C
Storage Temperature: Ambient
MDL Number: MFCD00006740
CAS Number: 119-91-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR