2,2'-Bipyrimidyl 97%
Supplier: Ambeed
|
Warning
|
A108261-100MG
A108261-10G
A108261-250MG
A108261-1G
103797-318EA
45
USD
103797-318
103823-098
77245-552
77245-554
77245-556
77309-942
2,2'-Bipyrimidyl 97%
2,2'-Bipyrimidyl
Precaución: for Research Use Only
Formula:
C₈H₆N₄ Boiling Pt: 395.4 °C (760 mm Hg) Melting Pt: 112…116 °C Flash Pt: 197 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014600 CAS Number: 34671-83-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...