2,2'-Bipyridine-6,6'-dicarboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2,2-Bipyridine-6,6-dicarboxylic Acid

A637367-1G A637367-5G A637367-250MG
103799-706EA 26 USD
103799-706 103840-320 76844-490 76844-492 77018-254
2,2'-Bipyridine-6,6'-dicarboxylic acid 97%
2,2'-Bipyridine-6,6'-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₈N₂O₄
MW: 244.21 g/mol
Boiling Pt: 568.083 °C (760 mm Hg)
Melting Pt: 286 °C
Flash Pt: 297.366 °C
Storage Temperature: Freezer
MDL Number: MFCD11042476
CAS Number: 4479-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR