2,2':5',2''-Terthiophene 98%
Supplier: Ambeed
|
Warning
|
A107281-10G
A107281-250MG
A107281-25G
A107281-1G
103797-158EA
6
USD
103797-158
103822-924
77245-498
77245-500
77245-502
77283-560
2,2':5',2''-Terthiophene 98%
2,2':5',2''-Terthiophene
Precaución: for Research Use Only
Formula:
C₁₂H₈S₃ MW: 248.39 g/mol Boiling Pt: 361.3 °C (760 mm Hg) Melting Pt: 93…95 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012167 CAS Number: 1081-34-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...