2,2,6,6-Tetramethylpiperidine 98%

Supplier: Ambeed
Danger

A706132-25G A706132-10G A706132-5G
103810-528EA 113 USD
103810-528 76844-876 77117-944 77117-946
2,2,6,6-Tetramethylpiperidine 98%
2,2,6,6-Tetramethylpiperidine

Precaución: for Research Use Only

Formula: C₉H₁₉N
MW: 141.26 g/mol
Boiling Pt: 152
Flash Pt: 24 °C
Storage Temperature: Ambient
MDL Number: MFCD00005985
CAS Number: 768-66-1
UN: 1992
ADR: 3,6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR