2,2,6,6-Tetramethyl-3,5-heptanedione 97%

Supplier: Ambeed
Warning

Synonyms: Dipivaloylmethane, TMHD, 2,2,6,6-Tetramethylheptane-3,5-dione

A622650-25G A622650-100G A622650-10G A622650-500G A622650-5G
103840-224EA 9 USD
103840-224 76844-390 77254-418 77254-420 77254-422 77307-980
2,2,6,6-Tetramethyl-3,5-heptanedione 97%
2,2,6,6-Tetramethyl-3,5-heptanedione

Precaución: for Research Use Only

Formula: C₁₁H₂₀O₂
MW: 184.28 g/mol
Boiling Pt: 72…73 °C (6 mm Hg)
Flash Pt: 67.22 °C
Storage Temperature: Ambient
MDL Number: MFCD00008848
CAS Number: 1118-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR