2,2,4-Trimethyl-1,2-dihydroquinoline 93%
Supplier: Ambeed
|
Warning
|
A280032-100G
A280032-250MG
A280032-5G
A280032-1G
A280032-25G
103813-492EA
10
USD
103813-492
103853-436
76723-256
76723-258
77093-366
77305-348
77305-350
2,2,4-Trimethyl-1,2-dihydroquinoline 93%
2,2,4-Trimethyl-1,2-dihydroquinoline
Precaución: for Research Use Only
Formula:
C₁₂H₁₅N Boiling Pt: 269.7±20.0 °C (760 mm Hg) Melting Pt: 120 °C Flash Pt: 118.6 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00044248 CAS Number: 147-47-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...