ω,2,4-Trichloroacetophenone 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
2,4-Dichlorophenacyl chloride, α,2,4-Trichloroacetophenone, 2,2',4'-Trichloroacetophenone
A863510-1000G
A863510-25G
A863510-100G
A863510-500G
103810-592EA
163
USD
103810-592
76845-522
76845-524
77256-382
77256-384
ω,2,4-Trichloroacetophenone 98%
ω,2,4-Trichloroacetophenone
Precaución: for Research Use Only
Formula:
C₈H₅Cl₃O MW: 223.49 g/mol Melting Pt: 47…54 °C Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000934 CAS Number: 4252-78-2 UN: 3265 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...