2,2,2-Trifluoro-3'-nitroacetophenone 97%

Supplier: Ambeed
Warning

A101277-250MG A101277-1G
103795-888EA 52 USD
103795-888 103821-574 77282-698 77297-042
2,2,2-Trifluoro-3'-nitroacetophenone 97%
2,2,2-Trifluoro-3'-nitroacetophenone

Precaución: for Research Use Only

Formula: C₈H₄F₃NO₃
Boiling Pt: 219.3 °C (760 mm Hg)
Flash Pt: 86.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00034541
CAS Number: 657-15-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR