2,2,2-Trifluoro-1-(3,4,5-trichlorophenyl)ethanone 95%

Supplier: Ambeed
Warning

A155307-100MG A155307-250MG A155307-1G
103804-742EA 42 USD
103804-742 103842-934 76984-800 76984-802 77300-108
2,2,2-Trifluoro-1-(3,4,5-trichlorophenyl)ethanone 95%
2,2,2-Trifluoro-1-(3,4,5-trichlorophenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₂Cl₃F₃O
MW: 277.45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25460357
CAS Number: 158401-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR